Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-propenamide, N-[2-hydroxy-2-(4-methoxyphenyl)ethyl]-3-phenyl-, (2E)- | 456-12-2 | MFCD00048045 | 99.8% | 297.35 | C18H19NO3 | 10 G
SDP

Catalog No. 5000209224
Encompass_Preferred

Aegeline is a high-purity natural alkaloid reference standard intended for analytical and pharmacological research. It is supplied as a solid and is also available as a DMSO solution for analytical use and method development.

  • Purity 99.81%
  • Cas number 456-12-2
  • Molecular formula C18H19NO3
  • Molecular weight 297.35 g/mol
  • Physical form Solid and DMSO solution
  • Typical applications Analytical standards, method development, and pharmacological research
  • Available package sizes 500 mg, 1 g, 5 g, 10 g, and 50 g (quote)
  • Product documents Data sheet and certificate of analysis available as PDF

Catalog No. 50-002-09224 Supplier Medchemexpress LLC Supplier No. HYW04215610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aegeline is a high-purity natural alkaloid reference standard intended for analytical and pharmacological research. It is supplied as a solid and is also available as a DMSO solution for analytical use and method development.

  • Purity 99.81%
  • Cas number 456-12-2
  • Molecular formula C18H19NO3
  • Molecular weight 297.35 g/mol
  • Physical form Solid and DMSO solution
  • Typical applications Analytical standards, method development, and pharmacological research
  • Available package sizes 500 mg, 1 g, 5 g, 10 g, and 50 g (quote)
  • Product documents Data sheet and certificate of analysis available as PDF

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.