Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Propenamide, N-[2-hydroxy-2-(4-methoxyphenyl)ethyl]-3-phenyl-, (2E)- | 456-12-2 | MFCD00048045 | 99.8% | 297.35 g/mol | C18H19NO3 | 1 G
SDP

Catalog No. 5000209428
Encompass_Preferred

Aegeline is an alkaloid reference standard intended for research use that mimics the yeast SNARE protein Sec22p and suppresses α-synuclein and Bax toxicity in yeast while exhibiting antioxidant activity.

  • Alkaloid reference standard for research use.
  • Cas number: 456-12-2.
  • Chemical formula: C18H19NO3; molecular weight: 297.35 g/mol.
  • Purity 99.81% (as provided on the certificate of analysis).
  • Form: white to off-white solid powder.
  • Solubility: soluble in DMSO (100 mg/mL); in vivo formulations listed for dosing preparations.
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solution at -80°C or -20°C for shorter periods.
  • Pack size: 1 g.

Catalog No. 50-002-09428 Supplier Medchemexpress LLC Supplier No. HYW0421561G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aegeline is an alkaloid reference standard intended for research use that mimics the yeast SNARE protein Sec22p and suppresses α-synuclein and Bax toxicity in yeast while exhibiting antioxidant activity.

  • Alkaloid reference standard for research use.
  • Cas number: 456-12-2.
  • Chemical formula: C18H19NO3; molecular weight: 297.35 g/mol.
  • Purity 99.81% (as provided on the certificate of analysis).
  • Form: white to off-white solid powder.
  • Solubility: soluble in DMSO (100 mg/mL); in vivo formulations listed for dosing preparations.
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solution at -80°C or -20°C for shorter periods.
  • Pack size: 1 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.