Learn More
Medchemexpress LLC 2-propenamide, N-[3-[2-[[4-[2-(dimethylamino)ethoxy]-2-methoxyphenyl]amino]...], trifluoroacetate | 2753348-63-7 | 99.4% | 628.60 | C30H31F3N6O6 | 10 MG
SDP
Catalog No. 5000297929
Encompass_Preferred
EGFR-IN-1 TFA is an orally active, irreversible small-molecule inhibitor selective for L858R/T790M mutant forms of the epidermal growth factor receptor (EGFR). It potently inhibits gefitinib-resistant EGFR mutants and demonstrates antiproliferative activity in mutant EGFR cell lines, making it suitable for cellular assays and preclinical research.
  • High mutant selectivity for L858R/T790M.
  • Irreversible mode of inhibition.
  • Potent antiproliferative activity in cell models.
  • Supplied as a trifluoroacetate salt for stability.
  • Intended for in vitro and preclinical research use.
Catalog No. 50-002-97929 Supplier Medchemexpress LLC Supplier No. HY19617B10MG
EGFR-IN-1 TFA is an orally active, irreversible small-molecule inhibitor selective for L858R/T790M mutant forms of the epidermal growth factor receptor (EGFR). It potently inhibits gefitinib-resistant EGFR mutants and demonstrates antiproliferative activity in mutant EGFR cell lines, making it suitable for cellular assays and preclinical research.
  • High mutant selectivity for L858R/T790M.
  • Irreversible mode of inhibition.
  • Potent antiproliferative activity in cell models.
  • Supplied as a trifluoroacetate salt for stability.
  • Intended for in vitro and preclinical research use.