Learn More
Medchemexpress LLC 2-Propenethioamide, 3-[3,5-bis(1,1-dimethylethyl)-4-hydroxyphenyl]-2-cyano-, (2E)- | 148741-30-4 | 99.3% | 316.46 | 25 MG
SDP
Catalog No. 5000402443
Encompass_Preferred
Tyrphostin AG 879 (AG 879) is a tyrosine kinase inhibitor that inhibits TrKA phosphorylation with an IC50 of 10 μM, but not TrKB and TrKC. It is also a selective ErbB2 tyrosine kinase inhibitor with an IC50 of 1 μM, showing at least 500-fold higher selectivity for ErbB2 over EGFR. This compound exhibits anticancer activity.
  • Significantly decreases cell proliferation in various cell lines
  • Induces a dose-dependent increase in apoptosis
  • Reduces tumor sizes in animal models
Catalog No. 50-004-02443 Supplier Medchemexpress LLC Supplier No. 25MGHY20878
Tyrphostin AG 879 (AG 879) is a tyrosine kinase inhibitor that inhibits TrKA phosphorylation with an IC50 of 10 μM, but not TrKB and TrKC. It is also a selective ErbB2 tyrosine kinase inhibitor with an IC50 of 1 μM, showing at least 500-fold higher selectivity for ErbB2 over EGFR. This compound exhibits anticancer activity.
  • Significantly decreases cell proliferation in various cell lines
  • Induces a dose-dependent increase in apoptosis
  • Reduces tumor sizes in animal models