Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-propenoic acid, 2-(bromomethyl)-, methyl ester | 4224-69-5 | MFCD00011697 | 99.4% | 179.01 | C5H7BrO2 | 1 ML
SDP

Catalog No. 5000303774
Encompass_Preferred

Methyl 2-(bromomethyl)acrylate is a brominated acrylate reagent used in chemical synthesis and biochemical research as an electrophilic alkylating and acrylate-building block. It is supplied as a high-purity material in solution and bulk liquid formats, with supporting documentation for quality and storage guidance.

  • High purity (99.38%) suitable for research applications.
  • Provided as 10 mM solution in DMSO and as bulk liquid sizes.
  • High solubility in DMSO (200 mg/mL); ultrasonic agitation recommended.
  • Storage: 4°C protected from light; in solvent store -80°C (6 months) or -20°C (1 month).
  • Molecular weight 179.01 and formula C5H7BrO2 for reference.
  • Datasheet and certificate of analysis available for batch-specific data.

Catalog No. 50-003-03774 Supplier Medchemexpress LLC Supplier No. HYW01658210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl 2-(bromomethyl)acrylate is a brominated acrylate reagent used in chemical synthesis and biochemical research as an electrophilic alkylating and acrylate-building block. It is supplied as a high-purity material in solution and bulk liquid formats, with supporting documentation for quality and storage guidance.

  • High purity (99.38%) suitable for research applications.
  • Provided as 10 mM solution in DMSO and as bulk liquid sizes.
  • High solubility in DMSO (200 mg/mL); ultrasonic agitation recommended.
  • Storage: 4°C protected from light; in solvent store -80°C (6 months) or -20°C (1 month).
  • Molecular weight 179.01 and formula C5H7BrO2 for reference.
  • Datasheet and certificate of analysis available for batch-specific data.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.