Learn More
Medchemexpress LLC 2-propenoic acid, 2-(bromomethyl)-, methyl ester | 4224-69-5 | MFCD00011697 | 99.4% | 179.01 | C5H7BrO2 | 10 G
SDP
Catalog No. 5000301833
Encompass_Preferred
Methyl 2-(bromomethyl)acrylate (CAS 4224-69-5) is a brominated methyl ester of acrylic acid used as a biochemical assay reagent and as a synthetic intermediate in organic and medicinal chemistry. Supplied for research applications, the material is accompanied by analytical documentation for traceability and handling guidance.
  • High purity of 99.38% by GC.
  • Available as neat material in multiple pack sizes and as a 10 mM solution in DMSO.
  • Provided with datasheet, certificate of analysis, and safety data sheet.
  • Suitable as a building block for synthesis and for assay development.
  • For research use only; not intended for human or clinical use.
  • Handle and store according to SDS instructions.
Catalog No. 50-003-01833 Supplier Medchemexpress LLC Supplier No. HYW01658210G
Methyl 2-(bromomethyl)acrylate (CAS 4224-69-5) is a brominated methyl ester of acrylic acid used as a biochemical assay reagent and as a synthetic intermediate in organic and medicinal chemistry. Supplied for research applications, the material is accompanied by analytical documentation for traceability and handling guidance.
  • High purity of 99.38% by GC.
  • Available as neat material in multiple pack sizes and as a 10 mM solution in DMSO.
  • Provided with datasheet, certificate of analysis, and safety data sheet.
  • Suitable as a building block for synthesis and for assay development.
  • For research use only; not intended for human or clinical use.
  • Handle and store according to SDS instructions.