Learn More
Medchemexpress LLC 2-Propenoic acid, 2-methyl-, 2-hydroxyethyl ester | 868-77-9 | 99.1% | 130.14 | 1 ML
SDP
Catalog No. 5000454866
Encompass_Preferred
2-Hydroxyethyl methacrylate (HEMA) is a monomer utilized in the creation of various polymers, and its polymer, PHEMA, is extensively employed in the synthesis of dental composite materials.
  • Monomer for various polymer syntheses.
  • Polymer (PHEMA) is widely used in dental composite materials.
Catalog No. 50-004-54866 Supplier Medchemexpress LLC Supplier No. HYW01272310MM/1ML
2-Hydroxyethyl methacrylate (HEMA) is a monomer utilized in the creation of various polymers, and its polymer, PHEMA, is extensively employed in the synthesis of dental composite materials.
  • Monomer for various polymer syntheses.
  • Polymer (PHEMA) is widely used in dental composite materials.