Learn More
Medchemexpress LLC 2-Propenoic acid, 3-(1H-imidazol-5-yl)-, (2E)- | 3465-72-3 | 99.8% | 138.13 | 1 ML
SDP
Catalog No. 5000440041
Encompass_Preferred
trans-urocanic acid (trans-UCA) is a natural epidermal constituent that inhibits human natural killer cell (NK) activity in vitro. It is active in regulating immune function.
  • Natural epidermal constituent.
  • Inhibits human natural killer cell (NK) activity in vitro.
  • Active in regulating an immune function.
  • Partially inhibits cytotoxic function of IL-2-activated NK cells and reduces IL-2-induced activation of NK cells.
  • Slightly decreases cell proliferation and viability of primary human keratinocytes at 100 μg/mL.
Catalog No. 50-004-40041 Supplier Medchemexpress LLC Supplier No. HY113008B10MM/1ML
trans-urocanic acid (trans-UCA) is a natural epidermal constituent that inhibits human natural killer cell (NK) activity in vitro. It is active in regulating immune function.
  • Natural epidermal constituent.
  • Inhibits human natural killer cell (NK) activity in vitro.
  • Active in regulating an immune function.
  • Partially inhibits cytotoxic function of IL-2-activated NK cells and reduces IL-2-induced activation of NK cells.
  • Slightly decreases cell proliferation and viability of primary human keratinocytes at 100 μg/mL.