Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-propenoic acid, 3-[2-[(1E)-2-(3,4-dihydroxyphenyl)ethenyl]-3,4-dihydroxyphenyl]-, (2E)- | 158732-59-3 | MFCD31560810 | 99.3% | 314.29 g/mol | C17H14O6 | 10MG
SDP

Catalog No. 5000236560
Encompass_Preferred

Salvianolic acid F is a polyphenolic natural product provided as a solid research reagent for biochemical and in vitro cancer research. It is commonly used in studies of cell signaling, anticancer activity, and KRAS-related pathways, and is supplied in small milligram pack sizes suitable for laboratory assays.

  • High purity: 99.3%.
  • Physical form: solid powder.
  • Molecular formula and weight: C17H14O6, 314.29 g/mol.
  • CAS number: 158732-59-3.
  • Suitable for in vitro biochemical and cell-based assays.
  • Available in small milligram quantities for research use.

Catalog No. 50-002-36560 Supplier Medchemexpress LLC Supplier No. HY12584710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Salvianolic acid F is a polyphenolic natural product provided as a solid research reagent for biochemical and in vitro cancer research. It is commonly used in studies of cell signaling, anticancer activity, and KRAS-related pathways, and is supplied in small milligram pack sizes suitable for laboratory assays.

  • High purity: 99.3%.
  • Physical form: solid powder.
  • Molecular formula and weight: C17H14O6, 314.29 g/mol.
  • CAS number: 158732-59-3.
  • Suitable for in vitro biochemical and cell-based assays.
  • Available in small milligram quantities for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.