Learn More
Medchemexpress LLC 2-Propenoic acid, 3-(2-methoxyphenyl)- | 6099-03-2 | 178.19 | 10 G
SDP
Catalog No. 5000415798
Encompass_Preferred
2-Methoxycinnamic acid is a noncompetitive inhibitor of tyrosinase. It is for research use only and not sold to patients.
  • Purity: 99.94%
  • Available in various sizes as solid or solution (10 mM in DMSO)
  • Derived from plants, specifically Ranunculaceae Pulsatilla chinensis (Bunge) Regel
  • Noncompetitive inhibitor of tyrosinase
  • Detailed documentation available: Data Sheet, COA, SDS, Handling Instructions, HNMR, RP-HPLC
Catalog No. 50-004-15798 Supplier Medchemexpress LLC Supplier No. HYN138610G
2-Methoxycinnamic acid is a noncompetitive inhibitor of tyrosinase. It is for research use only and not sold to patients.
  • Purity: 99.94%
  • Available in various sizes as solid or solution (10 mM in DMSO)
  • Derived from plants, specifically Ranunculaceae Pulsatilla chinensis (Bunge) Regel
  • Noncompetitive inhibitor of tyrosinase
  • Detailed documentation available: Data Sheet, COA, SDS, Handling Instructions, HNMR, RP-HPLC