Learn More
Medchemexpress LLC 2-Propenoic acid,3-(3-hydroxyphenyl)- | 588-30-7 | 100.0% | 164.16 | 100 MG
SDP
Catalog No. 5000436558
Encompass_Preferred
m-Coumaric acid is a polyphenol metabolite derived from caffeic acid, produced by gut microflora, and its concentration in human biofluids depends on diet. It is a BBB-penetrant metabolite of chlorogenic acid. In vitro, m-Coumaric acid stimulates cerebral nerves and can evoke neurite outgrowth in hippocampal neuronal cells, promoting neuronal differentiation.
  • Increases spontaneous locomotor activity in mice
  • Inhibits L-dopa oxidation by epidermis tyrosinase
  • Attenuates non-catalytic protein glycosylation in retinas of diabetic rats
  • Suitable for research and analytical applications
Catalog No. 50-004-36558 Supplier Medchemexpress LLC Supplier No. HY113357100MG
m-Coumaric acid is a polyphenol metabolite derived from caffeic acid, produced by gut microflora, and its concentration in human biofluids depends on diet. It is a BBB-penetrant metabolite of chlorogenic acid. In vitro, m-Coumaric acid stimulates cerebral nerves and can evoke neurite outgrowth in hippocampal neuronal cells, promoting neuronal differentiation.
  • Increases spontaneous locomotor activity in mice
  • Inhibits L-dopa oxidation by epidermis tyrosinase
  • Attenuates non-catalytic protein glycosylation in retinas of diabetic rats
  • Suitable for research and analytical applications