Learn More
Medchemexpress LLC 2-propenoic acid, 3-(4-hydroxy-3-methoxyphenyl)-, 1-methylethyl ester | 59831-94-6 | MFCD01736873 | 100.0% | 236.26 g/mol | C13H16O4 | 10 MG
SDP
Catalog No. 5000289823
Encompass_Preferred
Isopropyl ferulate is a small-molecule research chemical used as an antifungal and antioxidant agent. It is isolated from Notopterygium incisum and is commonly used in pharmaceutical research, cosmetic formulation studies, and as a reagent for synthesis and biological evaluation.
  • Used as an antifungal agent and antioxidant in research applications.
  • Suitable for pharmaceutical intermediate and formulation studies.
  • Reported high purity for reproducible experimental results.
  • Available in small, research-scale pack sizes for screening and method development.
  • Solid form with defined molecular formula and molecular weight for analytical work.
Catalog No. 50-002-89823 Supplier Medchemexpress LLC Supplier No. HYN420310MG
Isopropyl ferulate is a small-molecule research chemical used as an antifungal and antioxidant agent. It is isolated from Notopterygium incisum and is commonly used in pharmaceutical research, cosmetic formulation studies, and as a reagent for synthesis and biological evaluation.
  • Used as an antifungal agent and antioxidant in research applications.
  • Suitable for pharmaceutical intermediate and formulation studies.
  • Reported high purity for reproducible experimental results.
  • Available in small, research-scale pack sizes for screening and method development.
  • Solid form with defined molecular formula and molecular weight for analytical work.