Learn More
Medchemexpress LLC 2-propenoic acid, 3-(4-hydroxy-3-methoxyphenyl)-, 2-phenylethyl ester | 71835-85-3 | 99.7% | 298.33 | 5 MG
SDP
Catalog No. 5000453209
Encompass_Preferred
Phenethyl ferulate is a major constituent of Qianghuo. It demonstrates inhibitory activity against cyclooxygenase (COX) and 5-lipoxygenase (5-LOX) with IC50 values of 4.35 μM and 5.75 μM, respectively.
  • Inhibitory activity against cyclooxygenase (COX)
  • Inhibitory activity against 5-lipoxygenase (5-LOX)
Catalog No. 50-004-53209 Supplier Medchemexpress LLC Supplier No. HYW0092485MG
Phenethyl ferulate is a major constituent of Qianghuo. It demonstrates inhibitory activity against cyclooxygenase (COX) and 5-lipoxygenase (5-LOX) with IC50 values of 4.35 μM and 5.75 μM, respectively.
  • Inhibitory activity against cyclooxygenase (COX)
  • Inhibitory activity against 5-lipoxygenase (5-LOX)