Learn More
Medchemexpress LLC 2-Propenoic acid, 3-(4-hydroxyphenyl)-,(2E)- | 501-98-4 | 164.16 | 1 G
SDP
Catalog No. 5000389729
Encompass_Preferred
p-Coumaric acid (trans-4-Hydroxycinnamic acid) is an isomer of cinnamic acid with oral activity. It inhibits cell proliferation, promotes apoptosis, and exhibits antibacterial, anti-inflammatory, antioxidant, and anti-tumor activities. It is for research use only.
  • Inhibits cell proliferation.
  • Promotes apoptosis.
  • Exhibits antibacterial activity.
  • Exhibits anti-inflammatory activity.
  • Exhibits antioxidant activity.
  • Exhibits anti-tumor activities.
Catalog No. 50-003-89729 Supplier Medchemexpress LLC Supplier No. HYN03511G
p-Coumaric acid (trans-4-Hydroxycinnamic acid) is an isomer of cinnamic acid with oral activity. It inhibits cell proliferation, promotes apoptosis, and exhibits antibacterial, anti-inflammatory, antioxidant, and anti-tumor activities. It is for research use only.
  • Inhibits cell proliferation.
  • Promotes apoptosis.
  • Exhibits antibacterial activity.
  • Exhibits anti-inflammatory activity.
  • Exhibits antioxidant activity.
  • Exhibits anti-tumor activities.