Learn More
Medchemexpress LLC 2-propenoic acid, 3-phenyl-, butyl ester | 538-65-8 | MFCD00051560 | 99.7% | 204.27 g/mol | C13H16O2 | 10G
SDP
Catalog No. 5000239420
Encompass_Preferred
N-Butyl cinnamate is a cinnamic acid ester used as a biochemical reagent and organic intermediate for life-science research and organic synthesis. It is commonly used as a building block, fragrance/flavor intermediate, and reagent in biological studies. The product is supplied with supporting analytical documentation for research use.
Catalog No. 50-002-39420 Supplier Medchemexpress LLC Supplier No. HY12607010G
N-Butyl cinnamate is a cinnamic acid ester used as a biochemical reagent and organic intermediate for life-science research and organic synthesis. It is commonly used as a building block, fragrance/flavor intermediate, and reagent in biological studies. The product is supplied with supporting analytical documentation for research use.