Learn More
Medchemexpress LLC 2-propenoic acid, 3-phenyl-, ethyl ester, (2E)- | 4192-77-2 | MFCD00009189 | 98.5% | 176.21 g/mol | C11H12O2 | 500 G
SDP
Catalog No. 5000282259
Encompass_Preferred
(E)-Ethyl cinnamate is the ethyl ester of cinnamic acid supplied as a high-purity research reagent. It is a colorless to off-white liquid with molecular formula C11H12O2 and molecular weight 176.21 g/mol, commonly found in natural extracts such as davana oil and used as a fragrance component, flavoring intermediate, and chemical research material.
  • High purity (98.5%).
  • Colorless to off-white liquid form, density ~1.05 g/cm3.
  • Molecular formula C11H12O2; molecular weight 176.21 g/mol.
  • Suitable for use as a fragrance and flavor intermediate and as a research reagent.
  • Available in 500 G pack size for laboratory use.
  • Batch certificate of analysis provided.
Catalog No. 50-002-82259 Supplier Medchemexpress LLC Supplier No. HYW236273500G
(E)-Ethyl cinnamate is the ethyl ester of cinnamic acid supplied as a high-purity research reagent. It is a colorless to off-white liquid with molecular formula C11H12O2 and molecular weight 176.21 g/mol, commonly found in natural extracts such as davana oil and used as a fragrance component, flavoring intermediate, and chemical research material.
  • High purity (98.5%).
  • Colorless to off-white liquid form, density ~1.05 g/cm3.
  • Molecular formula C11H12O2; molecular weight 176.21 g/mol.
  • Suitable for use as a fragrance and flavor intermediate and as a research reagent.
  • Available in 500 G pack size for laboratory use.
  • Batch certificate of analysis provided.