Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-propenoic acid, homopolymer | 9003-01-4 | MFCD00084394 | 3,000,000 (average) | (C3H4O2)x | 50 G
SDP

Catalog No. 5000280818
Encompass_Preferred

Poly(acrylic acid) (average Mw 3,000,000) is a high-molecular-weight anionic polymer supplied as a white to off-white solid for research and materials applications. It is used in formulations, coatings, and studies requiring polyelectrolytes or corrosion inhibition, and is provided in laboratory-scale pack sizes for ease of use.

  • High molecular weight (average Mw 3,000,000).
  • White to off-white solid powder.
  • Anionic polymer useful as a corrosion inhibitor and surface stabilizer.
  • Chemical formula: (C3H4O2)x; CAS number: 9003-01-4.
  • Available in multiple pack sizes for lab use.
  • Store at room temperature; solution storage recommended at lower temperatures.

Catalog No. 50-002-80818 Supplier Medchemexpress LLC Supplier No. HYW243303B50G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Poly(acrylic acid) (average Mw 3,000,000) is a high-molecular-weight anionic polymer supplied as a white to off-white solid for research and materials applications. It is used in formulations, coatings, and studies requiring polyelectrolytes or corrosion inhibition, and is provided in laboratory-scale pack sizes for ease of use.

  • High molecular weight (average Mw 3,000,000).
  • White to off-white solid powder.
  • Anionic polymer useful as a corrosion inhibitor and surface stabilizer.
  • Chemical formula: (C3H4O2)x; CAS number: 9003-01-4.
  • Available in multiple pack sizes for lab use.
  • Store at room temperature; solution storage recommended at lower temperatures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.