Learn More
Medchemexpress LLC 2-propenoic acid, homopolymer | 9003-01-4 | MFCD00084394 | 4,000,000 (average) | (C3H4O2)x | 5 G
SDP
Catalog No. 5000280813
Encompass_Preferred
Poly(acrylic acid) (MW 4,000,000) is a high-molecular-weight anionic polymer supplied as a white to off-white solid for research applications. It is used in biochemical assays and as a corrosion inhibitor and surface stabilizer; preparation and storage guidance are provided in the product datasheet.
  • High molecular weight (4,000,000 average)
  • White to off-white solid appearance
  • Water soluble; recommended 100 mg/mL in H2O with ultrasonic aid
  • Used as a corrosion inhibitor and surface stabilizer
  • Storage: room temperature for solid; solutions require low-temperature storage
Catalog No. 50-002-80813 Supplier Medchemexpress LLC Supplier No. HYW243303C5G
Poly(acrylic acid) (MW 4,000,000) is a high-molecular-weight anionic polymer supplied as a white to off-white solid for research applications. It is used in biochemical assays and as a corrosion inhibitor and surface stabilizer; preparation and storage guidance are provided in the product datasheet.
  • High molecular weight (4,000,000 average)
  • White to off-white solid appearance
  • Water soluble; recommended 100 mg/mL in H2O with ultrasonic aid
  • Used as a corrosion inhibitor and surface stabilizer
  • Storage: room temperature for solid; solutions require low-temperature storage