Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-propoxyaniline

Catalog No. CDS014846 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS014846 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS014846 Supplier Sigma Aldrich Supplier No. CDS014846100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-bromo-1,1,1-trifluoroacetone reacts with potassium enolate of ethyl 4,4,4-trifluoroacetoacetate to yield ethyl 2,4-bis(trifluoromethyl)-4-hydroxydihydro-3-furoate.3-bromo-1,1,1-trifluoroacetone may be used in the synthesis of:
  • 3-nonylthio-1,1,1-trifluoropropan-2-one
  • Cyclic tetrapeptides containing trifluoromethyl and pentafluoroethyl ketone as zinc binding functional group
  • Perfluoroalkylated trans-allylic alcohols

Specifications

CAS 4469-78-7
Molecular Formula C9H13NO
Linear Formula C9H13NO
MDL Number MFCD00628992
Quantity 5 g
Molecular Weight (g/mol) 151.21
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.