Learn More
AdipoGen 2-Propyl-2-pentenoic acid
SDP
Catalog No. 502560222
Encompass_Preferred
Chemical. CAS 60218-41-9. Formula C8H14O2. MW 142.2. Synthetic A major beta-oxidation metabolite of valproic acid VPA. Exhibits anticonvulsive potency similar to VPA and may be at least partially responsible for its neurological activity. Compound can be used as analytical reference material.
Catalog No. 50-256-0222 Supplier AdipoGen Supplier No. CDXP0155M025
Chemical. CAS 60218-41-9. Formula C8H14O2. MW 142.2. Synthetic A major beta-oxidation metabolite of valproic acid VPA. Exhibits anticonvulsive potency similar to VPA and may be at least partially responsible for its neurological activity. Compound can be used as analytical reference material.