Learn More
Medchemexpress LLC (±)-2-propyl-4-pentenoic acid | 1575-72-0 | MFCD00869309 | 98.0% | 142.20 g/mol | C8H14O2 | 10MG
SDP
Catalog No. 5000239012
Encompass_Preferred
Racemic 2-Propyl-4-pentenoic acid (4-en-VPA) is a research-grade analytical standard of a major toxic metabolite of valproic acid, used in studies of metabolism and developmental neurotoxicity. Supplied as a high-purity chemical, it is intended for analytical and in vitro research applications and is not for human or clinical use.
  • High purity: 98.0%.
  • Molecular formula C8H14O2; molecular weight 142.20 g/mol.
  • Available in small analytical sizes: 10 MG, 25 MG, 50 MG, 100 MG, 200 MG.
  • Suitable as an analytical standard for metabolic and toxicology studies.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-39012 Supplier Medchemexpress LLC Supplier No. HY12408710MG
Racemic 2-Propyl-4-pentenoic acid (4-en-VPA) is a research-grade analytical standard of a major toxic metabolite of valproic acid, used in studies of metabolism and developmental neurotoxicity. Supplied as a high-purity chemical, it is intended for analytical and in vitro research applications and is not for human or clinical use.
  • High purity: 98.0%.
  • Molecular formula C8H14O2; molecular weight 142.20 g/mol.
  • Available in small analytical sizes: 10 MG, 25 MG, 50 MG, 100 MG, 200 MG.
  • Suitable as an analytical standard for metabolic and toxicology studies.
  • For research use only; not for human or clinical use.