Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propylcyclohexanone 98.0+%, TCI America™
SDP

Catalog No. P111410ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P111410ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P111410ML Supplier TCI America Supplier No. P111410ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 94-65-5
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.23
MDL Number MFCD00019464
InChI Key OCJLPZCBZSCVCO-UHFFFAOYNA-N
PubChem CID 7199
IUPAC Name 2-propylcyclohexan-1-one
SMILES CCCC1CCCCC1=O

Specifications

Color Yellow
Boiling Point 198°C
Quantity 10 mL
Formula Weight 140.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Propylcyclohexanone
EINECSNumber (3)-2411
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.