Learn More
Medchemexpress LLC 2-propylpentanoic acid | 99-66-1 | MFCD00002672 | 100.0% | 144.21 g/mol | C8H16O2 | 100 MG
SDP
Catalog No. 5000241866
Encompass_Preferred
Valproic acid (2-propylpentanoic acid) analytical standard for research and analytical applications. It is provided as a high-purity reference material for qualitative and quantitative assays and for method development in HPLC, GC, and MS. A certificate of analysis is available and storage should follow COA instructions.
  • High purity analytical standard (99.97%).
  • Intended for HPLC, GC, and MS method development.
  • Suitable for qualitative and quantitative assays.
  • Certificate of analysis available for lot-specific data.
  • Chemical formula C8H16O2; molecular weight 144.21 g/mol.
  • Supplied as 100 mg quantity.
Catalog No. 50-002-41866 Supplier Medchemexpress LLC Supplier No. HY10585R100MG
Add to Cart
Add to Cart
Valproic acid (2-propylpentanoic acid) analytical standard for research and analytical applications. It is provided as a high-purity reference material for qualitative and quantitative assays and for method development in HPLC, GC, and MS. A certificate of analysis is available and storage should follow COA instructions.
  • High purity analytical standard (99.97%).
  • Intended for HPLC, GC, and MS method development.
  • Suitable for qualitative and quantitative assays.
  • Certificate of analysis available for lot-specific data.
  • Chemical formula C8H16O2; molecular weight 144.21 g/mol.
  • Supplied as 100 mg quantity.