Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propylphenol 98.0+%, TCI America™
SDP

Catalog No. P053025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P053025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P053025ML Supplier TCI America Supplier No. P053025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 644-35-9
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002251
InChI Key LCHYEKKJCUJAKN-UHFFFAOYSA-N
Synonym 2-n-propylphenol, o-propylphenol, phenol, 2-propyl, phenol, o-propyl, 1-hydroxy-2-n-propylbenzene, 1-2-hydroxyphenyl propane, propylphenol, 2-propyl-phenol, o-n-propylphenol, unii-333r6f6t3n
PubChem CID 12570
IUPAC Name 2-propylphenol
SMILES CCCC1=CC=CC=C1O

Specifications

Color Yellow
Boiling Point 214°C
Quantity 25 mL
UN Number 3145
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Propylphenol
EINECSNumber (4)-0057
RTECSNumber SM8600000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.