Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propylthiazole 98.0+%, TCI America™
SDP

Catalog No. P18375G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P18375G Supplier TCI America Supplier No. P18375G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 17626-75-4
Color Yellow
Boiling Point 172°C
Molecular Formula C6H9NS
Quantity 5 g
MDL Number MFCD00053154
UN Number 1993
Synonym 2-propylthiazole, thiazole, 2-propyl, 2-n-propylthiazole, unii-t395979vrl, 2-propylthiazol, thiazole,2-propyl, 2-normal-propyl thiazole, acmc-209eb6, #
InChI Key CMOIEFFAOUQJPS-UHFFFAOYSA-N
SMILES CCCC1=NC=CS1
IUPAC Name 2-propyl-1,3-thiazole
Molecular Weight (g/mol) 127.205
PubChem CID 87198
Formula Weight 127.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Propylthiazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.