Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Propylthio)ethanol 98.0+%, TCI America™
SDP

Catalog No. P07345ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P07345ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P07345ML Supplier TCI America Supplier No. P07345ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22812-90-4
Molecular Formula C5H12OS
Molecular Weight (g/mol) 120.21
MDL Number MFCD00014041
InChI Key KCWWXXYQPUDKBX-UHFFFAOYSA-N
Synonym 2-propylthio ethanol, 2-hydroxyethyl propyl sulfide, 2-n-propylthio ethanol, 2-hydroxyethyl n-propyl sulfide, 2-propylsulfanyl ethanol, 2-propylthioethan-1-ol, acmc-209fyz, ethanol, 2-propylthio, 2-propylsulfanyl ethanol #, 2-hydroxyethyln-propylsulfide
PubChem CID 89850
IUPAC Name 2-propylsulfanylethanol
SMILES CCCSCCO

Specifications

Color Yellow
Boiling Point 94°C
Quantity 5 mL
Formula Weight 120.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Propylthio)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.