Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Propynyl methanesulfonate

Catalog No. 80999325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-999-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 80-999-325G Supplier Sigma Aldrich Supplier No. 80999325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The presence of electrolyte additives enhances the performance of lithium ion batteries by improving their thermal stability and ionic conductivity. Additives also reduce the flammability of electrolytes and provide overcharge protection. 2-Propynyl methanesulfonate finds application as additive in lithium ion batteries.Product of MU Ionic Solutions Corp

Specifications

CAS 16156-58-4
Molecular Formula C4H6O3S
Linear Formula C4H6O3S
Quantity 25 g
MDL Number MFCD09841205
Synonym 2-Propinyl methanesulfonate; Methanesulfonic acid
Molecular Weight (g/mol) 134.15
Percent Purity ≥99.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.