Learn More
Medchemexpress LLC 2-Pyrazinecarboxamide, 3-amino-6-[4-(methylsulfonyl)phenyl]-N-phenyl- | 1232410-49-9 | 99.7% | 368.41 | 10 MG
SDP
Catalog No. 5000365548
Encompass_Preferred
VE-821 is a potent ATP-competitive inhibitor of ATR with Ki/IC50 of 13 nM/26 nM. It demonstrates excellent selectivity for ATR with minimal cross-reactivity against related PIKKs such as ATM, DNA-PK, mTOR, and PI3Kγ. This compound enhances the sensitivity of pancreatic cancer cells to radiation and Gemcitabine, and inhibits radiation-induced G2/M arrest in cancer cells.
  • Potent ATP-competitive inhibitor of ATR
  • Excellent selectivity for ATR with minimal cross-reactivity
  • Enhances sensitivity of pancreatic cancer cells to radiation and Gemcitabine
  • Inhibits radiation-induced G2/M arrest in cancer cells
  • Inhibits phosphorylation of Chk1 (Ser 345)
Catalog No. 50-003-65548 Supplier Medchemexpress LLC Supplier No. 10MGHY14731
VE-821 is a potent ATP-competitive inhibitor of ATR with Ki/IC50 of 13 nM/26 nM. It demonstrates excellent selectivity for ATR with minimal cross-reactivity against related PIKKs such as ATM, DNA-PK, mTOR, and PI3Kγ. This compound enhances the sensitivity of pancreatic cancer cells to radiation and Gemcitabine, and inhibits radiation-induced G2/M arrest in cancer cells.
  • Potent ATP-competitive inhibitor of ATR
  • Excellent selectivity for ATR with minimal cross-reactivity
  • Enhances sensitivity of pancreatic cancer cells to radiation and Gemcitabine
  • Inhibits radiation-induced G2/M arrest in cancer cells
  • Inhibits phosphorylation of Chk1 (Ser 345)