Learn More
Medchemexpress LLC 2-pyrazinecarboxamide, 3-amino-N-(aminoiminomethyl)-5-(dimethylamino)-6-(2-phenylethynyl)-hydrochloride | 2237925-62-9 | 98.0% | C16H18ClN7O | 10MG
SDP
Catalog No. 5000260099
Encompass_Preferred
DMA-135 hydrochloride is the hydrochloride salt of a small-molecule ligand that targets the enterovirus 71 (EV71) SLII RNA domain and inhibits EV71 IRES-dependent translation and replication; it has a reported KD of ~520 nM and shows no significant toxicity in cell-based studies. Provided as a light-yellow solid with high purity for biochemical and cellular research.
  • Binds EV71 SLII RNA with moderate affinity.
  • Inhibits IRES-dependent translation and viral replication in vitro.
  • High purity suitable for biochemical and cell-based assays.
  • Available as solid and as 10 mM solution in DMSO.
  • Light yellow solid appearance for easy visual inspection.
  • Stable when stored sealed; follow recommended temperature guidelines.
Catalog No. 50-002-60099 Supplier Medchemexpress LLC Supplier No. HY14593210MG
DMA-135 hydrochloride is the hydrochloride salt of a small-molecule ligand that targets the enterovirus 71 (EV71) SLII RNA domain and inhibits EV71 IRES-dependent translation and replication; it has a reported KD of ~520 nM and shows no significant toxicity in cell-based studies. Provided as a light-yellow solid with high purity for biochemical and cellular research.
  • Binds EV71 SLII RNA with moderate affinity.
  • Inhibits IRES-dependent translation and viral replication in vitro.
  • High purity suitable for biochemical and cell-based assays.
  • Available as solid and as 10 mM solution in DMSO.
  • Light yellow solid appearance for easy visual inspection.
  • Stable when stored sealed; follow recommended temperature guidelines.