Learn More
Medchemexpress LLC 2-Pyrazinecarboxamide, 6-ethyl-5-[(trans-4-methoxycyclohexyl)amino]-3-[[3-methoxy-4-[4-(4-methyl-1-piperazinyl)-1-piperi | 1373409-08-5 | 98.02% | 580.76 | 1 ML
SDP
Catalog No. 5000439340
Encompass_Preferred
EML4-ALK kinase inhibitor 1 is a potent, orally active inhibitor of echinoderm microtubule-associated protein-like 4-anaplastic lymphoma kinase (EML4-ALK), exhibiting an IC50 of 1 nM. This product is intended for research use only.
  • Potent orally active inhibitor of EML4-ALK with an IC50 of 1 nM.
  • Appears as a solid with a light yellow to yellow color.
Catalog No. 50-004-39340 Supplier Medchemexpress LLC Supplier No. HY11175210MM/1ML
EML4-ALK kinase inhibitor 1 is a potent, orally active inhibitor of echinoderm microtubule-associated protein-like 4-anaplastic lymphoma kinase (EML4-ALK), exhibiting an IC50 of 1 nM. This product is intended for research use only.
  • Potent orally active inhibitor of EML4-ALK with an IC50 of 1 nM.
  • Appears as a solid with a light yellow to yellow color.