Learn More
Medchemexpress LLC 2-Pyrazinecarboxylic acid | 98-97-5 | 99.5% | C5H4N2O2 | 1 ML
SDP
Catalog No. 5000434518
Encompass_Preferred
2-Pyrazinecarboxylic acid is a biochemical reagent suitable for biological material and organic compound research in life science. It acts as an active metabolite of pyrazinamide, an anti-tubercular agent. In vitro studies have shown its involvement in urate uptake and suppression of 1-aminocyclopropane-1-carboxylic acid oxidase (ACO) activity in plant cells, an enzyme crucial for ethylene formation.
  • Biochemical reagent for life science research
  • Active metabolite of pyrazinamide
  • Involved in urate uptake regulation
  • Suppresses ACO activity in plant cells
  • For research use only, not for medical applications
Catalog No. 50-004-34518 Supplier Medchemexpress LLC Supplier No. HY7635310MM/1ML
2-Pyrazinecarboxylic acid is a biochemical reagent suitable for biological material and organic compound research in life science. It acts as an active metabolite of pyrazinamide, an anti-tubercular agent. In vitro studies have shown its involvement in urate uptake and suppression of 1-aminocyclopropane-1-carboxylic acid oxidase (ACO) activity in plant cells, an enzyme crucial for ethylene formation.
  • Biochemical reagent for life science research
  • Active metabolite of pyrazinamide
  • Involved in urate uptake regulation
  • Suppresses ACO activity in plant cells
  • For research use only, not for medical applications