Learn More
Medchemexpress LLC 2-Pyrazinecarboxylic acid | 98-97-5 | 99.5% | C5H4N2O2 | 25 G
SDP
Catalog No. 5000358455
Encompass_Preferred
2-Pyrazinecarboxylic acid is a biochemical reagent used as a biological material or organic compound for life science related research. It is an active metabolite of pyrazinamide, an anti-tubercular agent, and has been studied in contexts such as uricosuric agent evaluation and ethylene formation inhibition in plant cells.
  • Used as a biological material or organic compound for life science related research
  • Active metabolite of pyrazinamide, an anti-tubercular agent
  • Decreased urinary excretion in models evaluating uricosuric agents
  • Increases urate uptake in rat renal brush-border membrane vesicles
  • Suppresses activity of ACO, an enzyme in ethylene formation
Catalog No. 50-003-58455 Supplier Medchemexpress LLC Supplier No. 25GHY76353
2-Pyrazinecarboxylic acid is a biochemical reagent used as a biological material or organic compound for life science related research. It is an active metabolite of pyrazinamide, an anti-tubercular agent, and has been studied in contexts such as uricosuric agent evaluation and ethylene formation inhibition in plant cells.
  • Used as a biological material or organic compound for life science related research
  • Active metabolite of pyrazinamide, an anti-tubercular agent
  • Decreased urinary excretion in models evaluating uricosuric agents
  • Increases urate uptake in rat renal brush-border membrane vesicles
  • Suppresses activity of ACO, an enzyme in ethylene formation