Learn More
Medchemexpress LLC 2-Pyrazinecarboxylic acid | 98-97-5 | 99.52% | C5H4N2O2 | 25 MG
SDP
Catalog No. 5000357982
Encompass_Preferred
2-Pyrazinecarboxylic acid is a biochemical reagent suitable for life science related research. It functions as an active metabolite of pyrazinamide, an anti-tubercular agent, and has demonstrated roles in urate uptake and ethylene formation pathways.
  • Biochemical reagent for life science research
  • Active metabolite of pyrazinamide
  • Suppresses activity of 1-aminocyclopropane-1-carboxylic acid oxidase (ACO)
  • Increases urate uptake in renal brush-border membrane vesicles
  • Shows inverse correlation with urinary urate excretion
Catalog No. 50-003-57982 Supplier Medchemexpress LLC Supplier No. 25MGHY76353R
2-Pyrazinecarboxylic acid is a biochemical reagent suitable for life science related research. It functions as an active metabolite of pyrazinamide, an anti-tubercular agent, and has demonstrated roles in urate uptake and ethylene formation pathways.
  • Biochemical reagent for life science research
  • Active metabolite of pyrazinamide
  • Suppresses activity of 1-aminocyclopropane-1-carboxylic acid oxidase (ACO)
  • Increases urate uptake in renal brush-border membrane vesicles
  • Shows inverse correlation with urinary urate excretion