Learn More
Medchemexpress LLC 2-Pyrazinecarboxylic acid | 98-97-5 | 99.7% | C5H4N2O2 | 50 MG
SDP
Catalog No. 5000434645
Encompass_Preferred
2-Pyrazinecarboxylic acid (Standard) is the analytical standard of 2-Pyrazinecarboxylic acid. This product is intended for research and analytical applications. It is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Analytical standard grade, providing a reference standard assay
  • Suitable for qualitative, quantitative, and methodological research experiments
  • Commonly utilized in HPLC, GC, and MS applications
Catalog No. 50-004-34645 Supplier Medchemexpress LLC Supplier No. HY76353R50MG
2-Pyrazinecarboxylic acid (Standard) is the analytical standard of 2-Pyrazinecarboxylic acid. This product is intended for research and analytical applications. It is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Analytical standard grade, providing a reference standard assay
  • Suitable for qualitative, quantitative, and methodological research experiments
  • Commonly utilized in HPLC, GC, and MS applications