Learn More
Medchemexpress LLC 2-Pyrazinecarboxylic acid (standard) | 98-97-5 | 99.7% | C5H4N2O2 | 100 MG
SDP
Catalog No. 5000434103
Encompass_Preferred
2-Pyrazinecarboxylic acid (Standard) is an analytical standard and biochemical reagent intended for research and analytical applications. It can be used as a biological material or organic compound for life science related research.
  • Used as a reference standard in assays
  • Suitable for qualitative research experiments
  • Suitable for quantitative research experiments
  • Suitable for methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-34103 Supplier Medchemexpress LLC Supplier No. HY76353R100MG
2-Pyrazinecarboxylic acid (Standard) is an analytical standard and biochemical reagent intended for research and analytical applications. It can be used as a biological material or organic compound for life science related research.
  • Used as a reference standard in assays
  • Suitable for qualitative research experiments
  • Suitable for quantitative research experiments
  • Suitable for methodological research experiments in HPLC, GC, and MS