Learn More
Medchemexpress LLC 2-pyridin-2-yl-4,5-dihydro-3H-benzo[e]indazol-1-one | 1062648-63-8 | 99.9% | 263.29 g/mol | C16H13N3O | 5MG

Supplier: Medchemexpress LLC HY1263025MG
DMT1 blocker 2 (2-pyridin-2-yl-4,5-dihydro-3H-benzo[e]indazol-1-one) is a small-molecule research compound that directly inhibits divalent metal transporter 1 (DMT1) with an in vitro IC50 of 0.83 μM. It blocks iron uptake by enterocytes in vivo and is supplied as a high-purity solid suitable for biochemical and cellular studies.
- Direct DMT1 inhibitor, IC50 = 0.83 μM.
- High purity (99.9%).
- Molecular formula C16H13N3O; molecular weight 263.29 g/mol.
- Soluble in DMSO for stock solution preparation.
- Suitable for biochemical assays and cellular studies of iron transport.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.