Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(Pyridin-2-yl)isopropyl amine

Catalog No. 802166500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-216-6500MG 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 80-216-6500MG Supplier Sigma Aldrich Supplier No. 802166500MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-(Pyridin-2-yl)isopropyl amine

Specifications

CAS 52568-28-2
Density 0.9800 g/mL (at 25°C)
Molecular Formula C8H12N2
Refractive Index n/D 1.516
Linear Formula C8H12N2
MDL Number MFCD08729302
Quantity 10 g
Synonym PIP; PIP Directing Group; Shi Auxilliary
Molecular Weight (g/mol) 136.19
Percent Purity 95% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.