Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pyridin-3-ylaniline, 95%, Thermo Scientific™

Catalog No. CC41814CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC41814CB 250 mg Amber Glass Bottle
CC41814DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC41814CB Supplier Thermo Scientific Chemicals Supplier No. CC41814CB
Add to Cart

Chemical Identifiers

CAS 177202-83-4
Molecular Formula C11H10N2
Molecular Weight (g/mol) 170.22
MDL Number MFCD08060516
InChI Key MTYHJHATMPCNLW-UHFFFAOYSA-N
Synonym 2-pyridin-3-yl aniline, 2-3-pyridyl aniline, 2-pyridin-3-yl-phenylamine, 2-pyridin-3-ylphenylamine, benzenamine, 2-3-pyridinyl, 2-3-pyridyl phenylamine, 3-o-aminophenyl pyridine, 3-2-aminophenyl pyridine
PubChem CID 459519
SMILES NC1=CC=CC=C1C1=CN=CC=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 2-pyridin-3-ylaniline
Formula Weight 170.21
Percent Purity 95%
Chemical Name or Material 2-Pyridin-3-ylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.