Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-pyridinamine, 3-bromo-5-fluoro- | 869557-43-7 | 99.9% | 191.003 | 10 G
SDP

Catalog No. 5000452497
Encompass_Preferred

2-Amino-3-bromo-5-fluoropyridine (3-Bromo-5-fluoro-2-pyridinamine) is a drug intermediate used for the synthesis of fluorinated pyridines. This product is for research use only and has not been fully validated for medical applications.

  • Appearance: Solid, white to light brown
  • Storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light)

Catalog No. 50-004-52497 Supplier Medchemexpress LLC Supplier No. HYW01002310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-3-bromo-5-fluoropyridine (3-Bromo-5-fluoro-2-pyridinamine) is a drug intermediate used for the synthesis of fluorinated pyridines. This product is for research use only and has not been fully validated for medical applications.

  • Appearance: Solid, white to light brown
  • Storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.