Learn More
Medchemexpress LLC 2-Pyridinamine, 4-bromo- | 84249-14-9 | 99.93% | 173.01 | 25 G
SDP
Catalog No. 5000402877
Encompass_Preferred
2-Amino-4-bromopyridine (4-Bromopyridine-2-amine) is an important intermediate used for the synthesis of other active compounds. Its derivative compounds can inhibit tumor cell proliferation by inducing G1 phase arrest and effectively act on wild-type ALK and drug-resistant mutants. This solid appears off-white to yellow.
  • This compound is instrumental in the creation of 2-aminopyridine derivatives.
  • Derivative compounds have demonstrated the ability to inhibit the proliferation of tumor cells by inducing G1 phase arrest.
  • These derivatives are also effective against both wild-type ALK and drug-resistant mutants.
  • It appears as a solid with an off-white to yellow color.
Catalog No. 50-004-02877 Supplier Medchemexpress LLC Supplier No. 25GHY41000
2-Amino-4-bromopyridine (4-Bromopyridine-2-amine) is an important intermediate used for the synthesis of other active compounds. Its derivative compounds can inhibit tumor cell proliferation by inducing G1 phase arrest and effectively act on wild-type ALK and drug-resistant mutants. This solid appears off-white to yellow.
  • This compound is instrumental in the creation of 2-aminopyridine derivatives.
  • Derivative compounds have demonstrated the ability to inhibit the proliferation of tumor cells by inducing G1 phase arrest.
  • These derivatives are also effective against both wild-type ALK and drug-resistant mutants.
  • It appears as a solid with an off-white to yellow color.