Learn More
Medchemexpress LLC 2-Pyridinamine, 5,6-bis(4-methoxy-3-methylphenyl)- | 2306039-66-5 | 99.8% | C21H22N2O2 | 100 MG
SDP
Catalog No. 5000356058
Encompass_Preferred
WSB1 Degrader 1 is a potent and orally active WSB1 (WD repeat and SOCS box-containing 1) degrader, exhibiting anticancer metastatic effects.
  • Induces WSB1 degradation in H1299-WSB1 cells.
  • Exhibits potent antimigration efficacy in KHOS and H460 cell lines.
  • Significantly inhibits cancer cell migration under normoxia and hypoxia in KHOS cells.
  • Elevates RhoGDI2 protein levels in KHOS cells under hypoxia.
  • Significantly inhibits wound-healing of H1299-WSB1 cells.
  • Blocks wound-healing capability of wild-type A2780 cells.
  • Effectively inhibits pulmonary metastasis of cancer cells in mice bearing highly metastatic 4T1 breast cancer cells.
  • Shows quick absorption and acceptable blood exposure in rats after oral or intraperitoneal dosing.
Catalog No. 50-003-56058 Supplier Medchemexpress LLC Supplier No. HY141482100MG
Add to Cart
Add to Cart
WSB1 Degrader 1 is a potent and orally active WSB1 (WD repeat and SOCS box-containing 1) degrader, exhibiting anticancer metastatic effects.
  • Induces WSB1 degradation in H1299-WSB1 cells.
  • Exhibits potent antimigration efficacy in KHOS and H460 cell lines.
  • Significantly inhibits cancer cell migration under normoxia and hypoxia in KHOS cells.
  • Elevates RhoGDI2 protein levels in KHOS cells under hypoxia.
  • Significantly inhibits wound-healing of H1299-WSB1 cells.
  • Blocks wound-healing capability of wild-type A2780 cells.
  • Effectively inhibits pulmonary metastasis of cancer cells in mice bearing highly metastatic 4T1 breast cancer cells.
  • Shows quick absorption and acceptable blood exposure in rats after oral or intraperitoneal dosing.