Learn More
Medchemexpress LLC 2-Pyridinecarbohydrazide | 1452-63-7 | 99.8% | 137.14 | 100 G
SDP
Catalog No. 5000452537
Encompass_Preferred
2-Pyridinecarbohydrazide is a chemical compound extensively used as a building block in various fields of organic synthesis. This product is intended for research use only.
  • Extensively used in various fields of synthesis.
  • For research use only.
  • Stable as powder at -20°C for 3 years or 4°C for 2 years.
  • Stable in solvent at -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-004-52537 Supplier Medchemexpress LLC Supplier No. HYW010341100G
2-Pyridinecarbohydrazide is a chemical compound extensively used as a building block in various fields of organic synthesis. This product is intended for research use only.
  • Extensively used in various fields of synthesis.
  • For research use only.
  • Stable as powder at -20°C for 3 years or 4°C for 2 years.
  • Stable in solvent at -80°C for 6 months or -20°C for 1 month.