Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-pyridinecarbonitrile, 5-[(4'-amino-5',8'-difluoro-spiroquinazoline)carbonyl] hydrochloride | 1781934-50-6 | 99.8% | 100 MG
SDP

Catalog No. 5000231924
Encompass_Preferred

This hydrochloride salt is a potent, selective inducible nitric oxide synthase (iNOS) inhibitor with reported IC50 = 37 nM. Supplied as a high-purity research reagent suitable for in vitro and in vivo studies of iNOS-mediated pathways and inflammatory responses.

  • High purity 99.8% as reported by manufacturer.
  • Molecular weight 418.83.
  • Chemical formula C19H17ClF2N6O.
  • Hydrochloride salt form for improved solubility.
  • Potent, selective iNOS inhibition (IC50 = 37 nM).
  • Suitable for pharmacology and inflammation research applications.

Catalog No. 50-002-31924 Supplier Medchemexpress LLC Supplier No. HY12122A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This hydrochloride salt is a potent, selective inducible nitric oxide synthase (iNOS) inhibitor with reported IC50 = 37 nM. Supplied as a high-purity research reagent suitable for in vitro and in vivo studies of iNOS-mediated pathways and inflammatory responses.

  • High purity 99.8% as reported by manufacturer.
  • Molecular weight 418.83.
  • Chemical formula C19H17ClF2N6O.
  • Hydrochloride salt form for improved solubility.
  • Potent, selective iNOS inhibition (IC50 = 37 nM).
  • Suitable for pharmacology and inflammation research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.