Learn More
Medchemexpress LLC 2-Pyridinecarbothioamide | 5346-38-3 | 138.19 | 50 G
SDP
Catalog No. 5000445896
Encompass_Preferred
2-Pyridinecarbothioamide is a chemical compound primarily used as a versatile building block in various synthetic applications. It can be utilized for the synthesis of a Zn2+ responsive fluorescent probe, making it valuable in research and laboratory settings. This product is intended for research use only.
  • Acts as a versatile building block for chemical synthesis
  • Can be used in the creation of Zn2+ responsive fluorescent probes
  • Suitable for various research and laboratory applications
  • High purity level of 99.94%
Catalog No. 50-004-45896 Supplier Medchemexpress LLC Supplier No. HYW00235650G
2-Pyridinecarbothioamide is a chemical compound primarily used as a versatile building block in various synthetic applications. It can be utilized for the synthesis of a Zn2+ responsive fluorescent probe, making it valuable in research and laboratory settings. This product is intended for research use only.
  • Acts as a versatile building block for chemical synthesis
  • Can be used in the creation of Zn2+ responsive fluorescent probes
  • Suitable for various research and laboratory applications
  • High purity level of 99.94%