Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-pyridinecarboxaldehyde, 6-bromo-3-fluoro | 885267-36-7 | MFCD07781224 | 99.9% | 204.00 g/mol | C6H3BrFNO | 100 G
SDP

Catalog No. 5000210930
Encompass_Preferred

6-Bromo-3-fluoropyridine-2-carbaldehyde is a halogenated pyridine aldehyde used as a synthetic intermediate in medicinal chemistry and organic synthesis.

  • CAS number: 885267-36-7.
  • Molecular formula: C6H3BrFNO.
  • Molecular weight: 204.00 g/mol.
  • Purity (GC): 99.92%.
  • Appearance: white to light yellow solid.
  • Typical pack size: 100 g.
  • Storage: -20°C sealed and away from moisture; in solvent -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-002-10930 Supplier Medchemexpress LLC Supplier No. HYW058423100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Bromo-3-fluoropyridine-2-carbaldehyde is a halogenated pyridine aldehyde used as a synthetic intermediate in medicinal chemistry and organic synthesis.

  • CAS number: 885267-36-7.
  • Molecular formula: C6H3BrFNO.
  • Molecular weight: 204.00 g/mol.
  • Purity (GC): 99.92%.
  • Appearance: white to light yellow solid.
  • Typical pack size: 100 g.
  • Storage: -20°C sealed and away from moisture; in solvent -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.