Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-pyridinecarboxaldehyde, 6-bromo-3-fluoro | 885267-36-7 | MFCD07781224 | 99.9% | 204.00 g/mol | C6H3BrFNO | 50 G
SDP

Catalog No. 5000210947
Encompass_Preferred

6-Bromo-3-fluoropyridine-2-carbaldehyde is a pyridine-2-carbaldehyde derivative used as a drug intermediate in the synthesis of pharmaceutical and research compounds. It is supplied at high reported purity and is suitable for introducing a formyl group onto a halogenated pyridine core.

  • Used as a drug intermediate for synthesis of active compounds.
  • High reported purity (99.9%).
  • Contains bromine and fluorine substituents on the pyridine ring.
  • Available in multiple pack sizes including 50 G.
  • Molecular formula C6H3BrFNO and molecular weight 204.00 g/mol.

Catalog No. 50-002-10947 Supplier Medchemexpress LLC Supplier No. HYW05842350G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Bromo-3-fluoropyridine-2-carbaldehyde is a pyridine-2-carbaldehyde derivative used as a drug intermediate in the synthesis of pharmaceutical and research compounds. It is supplied at high reported purity and is suitable for introducing a formyl group onto a halogenated pyridine core.

  • Used as a drug intermediate for synthesis of active compounds.
  • High reported purity (99.9%).
  • Contains bromine and fluorine substituents on the pyridine ring.
  • Available in multiple pack sizes including 50 G.
  • Molecular formula C6H3BrFNO and molecular weight 204.00 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.