Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Pyridinecarboxaldehyde

Catalog No. P62003100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P62003100G 100 g
P6200325G 25 g
P62003500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P62003100G Supplier Sigma Aldrich Supplier No. P62003100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-pyridinecarboxaldehyde has been used:
  • To synthesize chitosan based bifunctionalized adsorbent
  • In the aldol addition reaction with pyruvate in the presence of 2-keto-3-deoxy-6-phosphogluconate aldolase (kdpg) catalyst to form (s)-4-hydroxy-2-keto-4-(2α-pyridyl)butyrate
  • In the condensation reaction with (s)-()-α-methylbenzylamine and (r)-(+)-α-methylbenzylamine to synthesize two chiral (s)-()- and (r)-(+)schiff base compounds.it can also be combined with thiosemicarbazide to form 2-pyridinecarboxaldehyde thiosemicarbazone ligand (L) which can further complex with ni(ii) and cu(ii) salts (mx) to form [m(L)2x2] complexes

Specifications

CAS 1121-60-4
Density 1.126 g/mL (at 25°C (literature))
Boiling Point 181°C (lit.)
Molecular Formula C6H5NO
Refractive Index n20/D 1.536 (literature)
Linear Formula C6H5NO
MDL Number MFCD00006290
Quantity 100 g
Synonym Picolinaldehyde; Pyridine-2-aldehyde
Molecular Weight (g/mol) 107.11
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.