Learn More
Medchemexpress LLC 2-Pyridinecarboxamide | 1452-77-3 | 100.0% | 122.13 g/mol | C6H6N2O | 25 G
SDP
Catalog No. 5000211914
Encompass_Preferred
Picolinamide is a small-molecule research chemical reported to inhibit poly(ADP-ribose) synthetase (PARS) in nuclei from rat pancreatic islet cells. It is supplied as a high-purity solid intended for laboratory research; consult the safety data sheet for handling and storage.
  • Inhibitor of poly(ADP-ribose) synthetase in reported studies.
  • High purity suitable for research assays.
  • Available in multiple pack sizes for laboratory use.
  • Solid form with a molecular weight of 122.13 g/mol.
  • CAS number 1452-77-3 for unambiguous identification.
Catalog No. 50-002-11914 Supplier Medchemexpress LLC Supplier No. HY10102025G
Picolinamide is a small-molecule research chemical reported to inhibit poly(ADP-ribose) synthetase (PARS) in nuclei from rat pancreatic islet cells. It is supplied as a high-purity solid intended for laboratory research; consult the safety data sheet for handling and storage.
  • Inhibitor of poly(ADP-ribose) synthetase in reported studies.
  • High purity suitable for research assays.
  • Available in multiple pack sizes for laboratory use.
  • Solid form with a molecular weight of 122.13 g/mol.
  • CAS number 1452-77-3 for unambiguous identification.