Learn More
Medchemexpress LLC 2-pyridinecarboxamide, 5-[[(6-chloro-2-pyridinyl)methyl](1-oxo-2-propen-1-yl)amino]-N-phenyl | 2305052-86-0 | MFCD06761688 | 99.9% | C21H17ClN4O2 | 10MG
SDP
Catalog No. 5000259801
Encompass_Preferred
BPK-25 is an acrylamide-containing small-molecule research reagent that promotes degradation of nucleosome remodeling and deacetylation (NuRD) complex proteins and inhibits TMEM173 (STING) activation, resulting in modulation of NF-κB signaling. Supplied as a high-purity solid for in vitro and biochemical research; not intended for human use.
  • Promotes NuRD complex protein degradation by covalent protein engagement.
  • Inhibits STING (TMEM173) activation by the cyclic dinucleotide cGAMP.
  • High purity suitable for biochemical assays and in vitro studies.
  • Supplied as an off-white to light yellow solid for convenient handling.
  • Recommended storage: 4°C under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).
  • For research use only; not for clinical or human use.
Catalog No. 50-002-59801 Supplier Medchemexpress LLC Supplier No. HY14155010MG
BPK-25 is an acrylamide-containing small-molecule research reagent that promotes degradation of nucleosome remodeling and deacetylation (NuRD) complex proteins and inhibits TMEM173 (STING) activation, resulting in modulation of NF-κB signaling. Supplied as a high-purity solid for in vitro and biochemical research; not intended for human use.
  • Promotes NuRD complex protein degradation by covalent protein engagement.
  • Inhibits STING (TMEM173) activation by the cyclic dinucleotide cGAMP.
  • High purity suitable for biochemical assays and in vitro studies.
  • Supplied as an off-white to light yellow solid for convenient handling.
  • Recommended storage: 4°C under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).
  • For research use only; not for clinical or human use.