Learn More
Medchemexpress LLC 2-Pyridinecarboxylic acid, 2-(2-pyridinyl)hydrazide | 53995-62-3 | 99.9% | 214.22 | 100 MG
SDP
Catalog No. 5000351059
Encompass_Preferred
PFI-90 is a selective inhibitor of histone demethylase (KDM3B) which inhibits PAX3-FOXO1 action. It induces apoptosis and myogenic differentiation, leading to increased cell death, and has potential for antitumor activity. This product is for research use only.
  • Selective inhibitor of histone demethylase (KDM3B)
  • Induces apoptosis and myogenic differentiation
  • Exhibits dose-response in various cell lines (RH4, RH30, OSA-CL, TC-32)
  • Increases apoptosis in RH4 and SCMC cells at 3 μM concentration
  • Potential for antitumor activity
Catalog No. 50-003-51059 Supplier Medchemexpress LLC Supplier No. HY139348100MG
PFI-90 is a selective inhibitor of histone demethylase (KDM3B) which inhibits PAX3-FOXO1 action. It induces apoptosis and myogenic differentiation, leading to increased cell death, and has potential for antitumor activity. This product is for research use only.
  • Selective inhibitor of histone demethylase (KDM3B)
  • Induces apoptosis and myogenic differentiation
  • Exhibits dose-response in various cell lines (RH4, RH30, OSA-CL, TC-32)
  • Increases apoptosis in RH4 and SCMC cells at 3 μM concentration
  • Potential for antitumor activity